Tlačiť
ESD safety shoes, Velcro® fastening, S3, jo_Clean Strap Low, Jori
Dodávateľ: ELTEN
12681/36 12681/37 12681/38 12681/39 12681/40 12681/41 12681/42 12681/43 12681/44 12681/45 12681/46 12681/47 12681/48 12691/36 12691/37 12691/38 12691/39 12691/40 12691/41 12691/42 12691/43 12691/44 12691/45 12691/46 12691/47 12691/48
ELTN12681/36EA 71.4 EUR
ELTN12681/36 ELTN12681/37 ELTN12681/38 ELTN12681/39 ELTN12681/40 ELTN12681/41 ELTN12681/42 ELTN12681/43 ELTN12681/44 ELTN12681/45 ELTN12681/46 ELTN12681/47 ELTN12681/48 ELTN12691/36 ELTN12691/37 ELTN12691/38 ELTN12691/39 ELTN12691/40 ELTN12691/41 ELTN12691/42 ELTN12691/43 ELTN12691/44 ELTN12691/45 ELTN12691/46 ELTN12691/47 ELTN12691/48
ESD safety shoes, Velcro® fastening, S3, jo_Clean Strap Low, Jori
Obuv
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • Composite toe cap
  • Metal- and leather-free
  • MONO-PU sole jo_MONO
  • Available in white or black finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certifikácie: EN ISO 20345 S3 SRC, Form A.