Tlačiť
Tyvek® 1422A autoclavable wrappers
Dodávateľ: KEYSTONE CLEANROOM PRODUCTS
T2ASHEET06 T2ASHEET10 T2ASHEET14 T2ASHEET0303 T2ASHEET0404 T2ASHEET0606 T2ASHEET0612 T2ASHEET075075 T2ASHEET0808 T2ASHEET0909 T2ASHEET0911 T2ASHEET1212 T2ASHEET1515 T2ASHEET1818 T2ASHEET2020 T2ASHEET2424 T2ASHEET3030 T2ASHEET3636 T2ASHEET4040 T2ASHEET4080 T2ASHEET5454 T2ASHEET60140 T2ASHEET6060 T2ASHEET7272 T2ASHEET8494
129-3042EA 3616.93 EUR
129-3042 129-3049 129-3051 129-3039 129-3041 129-3043 129-3044 129-3045 129-3046 129-3047 129-3048 129-3050 129-3052 129-3053 129-3055 129-3056 129-2981 129-2984 129-2985 129-2986 129-2990 129-2991 129-2992 129-2993 129-2994
Tyvek® 1422A autoclavable wrappers
Utěrky sterilizační
Improve the microbial barrier and cleanliness of your sterilisation wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic and lint-free Tyvek® 1422A
Drapeable Tyvek® 1422A Wrappers are an improvement on cellulose or SMS based wrapping materials for steam sterilisation. Eliminate the wasted time and expense of cutting larger square sheets of sterilisation wrap to the desired sizes. Round shaped wrappers are an ideal way to cover round openings. Clean cut round and rectangular/square sheets are delivered in labelled, sealed poly bags to ensure cleanliness. Simply secure in place with autoclavable elastic bands.
Certifikácie: Certificates of Conformance sent with each shipment.
Pozor: Note: Dimensions shown are W×L.