Tlačiť
Magnetic stirrer heads, BOLA
Dodávateľ: Bohlender
C520-28 C520-24 C520-48
442-0759EA 1314.88 EUR
442-0759 442-0758 442-0760
Magnetic stirrer heads, BOLA
Hlavy míchadel
PTFE compound, glass shaft. These gastight stirrer heads are ideal for PTFE-jacketed stirrer shafts from BOLA and are also suitable for shortened stirrer shafts. Suitable for high speeds of up to 1500 min⁻¹.
  • Very good chemical resistance, temperature resistance from –200 to +250 °C
  • Joint-Cone with nut for easy locking and unlocking of the ground joint
  • Adjustable stirrer shaft hight ±40 mm
  • Compression fittings for safe fixing of stirrer shaft and optimum power transmission
  • No leakage or memory effects due to non-porous, welded rotor
Powerful transmission of up to 90 Ncm. The insertion length of shafts is 95 mm and the drive shaft has an outer Ø of 50 mm.
Powerful transmission of up to 90 Ncm. The insertion length of shafts is 95 mm and the drive shaft has an outer Ø of 50 mm.