Tlačiť
Multiple distributors for bottles with GL 45 thread, BOLA
Dodávateľ: Bohlender
D618-48 D618-16 D618-24 D618-44 D618-46
215-3895EA 204.69 EUR
215-3895 201-0296 215-3893 201-0295 215-3894
Multiple distributors for bottles with GL 45 thread, BOLA
Uzávěry Uzávěry k láhvím
PPS screw cap, black, distributor body made from PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with different external diameters can be inserted through the necks.