Tlačiť
Texture analysers, CT3
Dodávateľ: AMETEK
CT31000230 CT3100230 CT31500230 CT310K230 CT34500230
646-0000EA 0 EUR
646-0000 646-0001 646-0003 646-0002 646-0004
Texture analysers, CT3
Analyzátory Analyzátory Textury
The CT3 are robust analysers used for both compression and tensile testing in environments where routine tests are required.
  • Six built-in test modes with storage for up to ten customised test methods
  • Two adjustable base tables for versatility with position range of 1 to 100 mm for larger samples
  • A range of probes available for new application opportunities
It calculates a number of physical properties correlated to human sensory evaluation of food and other consumer products. Custom reports and graphs can be created using the optional software Texture Pro CT.
Certifikácie: GME and GMIA monographs for gelatine bloom assessment with AOAC probe included in TA-P-Kit2.
Informace o dodávce: Supplied with software and USB cable.