Tlačiť
Single element standards for ICP-OES, sodium
Dodávateľ: Agilent
5190-8454 5190-8525 5190-8526 5190-8206
HEWL5190-8454EA 155.64 EUR
HEWL5190-8454 HEWL5190-8525 HEWL5190-8526 HEWL5190-8206
Single element standards for ICP-OES, sodium
Standardy Standardy kationické Standardy - sodík
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.