Tlačiť
Single element standards for ICP-OES, tttrium
Dodávateľ: Agilent
5190-8555 5190-8556 5190-8232 5190-8233
HEWL5190-8555EA 90.65 EUR
HEWL5190-8555 HEWL5190-8556 HEWL5190-8232 HEWL5190-8233
Single element standards for ICP-OES, tttrium
Standardy Standardy kationické Standardy - yttrium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.