Tlačiť
Connectors, with GL thread
Dodávateľ: Bohlender
D538-18 D540-18 D539-18 D541-18 D539-14 D540-14 D541-14 D538-14 D539-25 D538-32 D539-32 D540-25 D538-25 D539-45 D538-45 D541-25 D540-32 D540-45
229-0010EA 29.36 EUR
229-0010 229-0055 229-0052 229-0258 229-0051 229-0054 229-0257 229-0000 229-0053 229-0251 229-0253 229-0056 229-0106 229-0254 229-0252 229-0259 229-0255 229-0256
Connectors, with GL thread
Fitinky a konektory
PTFE
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −200 to +250 °C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable
Connector with two, three or four GL screw connections. To distribute liquids or gases. Can be used as a reducer to connect different tubes or hoses.