Tlačiť
Multiple distributor for Jerrican, S thread with GL thread connectors, BOLA
Dodávateľ: Bohlender
D760-16 D760-24
201-0292EA 208.18 EUR
201-0292 201-0291
Multiple distributor for Jerrican, S thread with GL thread connectors, BOLA
Uzávěry Uzávěry k láhvím
PTFE.
  • Excellent chemical resistance, the medium only comes into contact with PTFE
  • Temperature resistant from –200 to +250 °C
  • Autoclavable (121 °C)
Screw cap made from glass fibre reinforced PTFE suitable for containers with S 55 or S 60 threads. Movable distributor body with GL connector necks made from PTFE. For the removal and filling of aggressive or pure liquids and insertion of hoses, tubes and probes into containers.