Tlačiť
Single element standards for ICP-OES, silicon
Dodávateľ: Agilent
5190-8450 5190-8451 5190-8521 5190-8522
HEWL5190-8450EA 194.95 EUR
HEWL5190-8450 HEWL5190-8451 HEWL5190-8521 HEWL5190-8522
Single element standards for ICP-OES, silicon
Standardy Standardy kationické Standardy - křemík
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.