Tlačiť
Single element standards for ICP-OES, tellurium
Dodávateľ: Agilent
5190-8533 5190-8534 5190-8213 5190-8214
HEWL5190-8533EA 135.58 EUR
HEWL5190-8533 HEWL5190-8534 HEWL5190-8213 HEWL5190-8214
Single element standards for ICP-OES, tellurium
Standardy Standardy kationické Standardy - tellur
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.