Tlačiť
Single element standards for ICP-OES, indium
Dodávateľ: Agilent
5190-8467 5190-8468 5190-8398 5190-8399
HEWL5190-8467EA 135.58 EUR
HEWL5190-8467 HEWL5190-8468 HEWL5190-8398 HEWL5190-8399
Single element standards for ICP-OES, indium
Standardy Standardy kationické Standardy - indium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.