Tlačiť
Single element standards for ICP-OES, germanium
Dodávateľ: Agilent
5190-8390 5190-8391
HEWL5190-8390EA 194.95 EUR
HEWL5190-8390 HEWL5190-8391
Single element standards for ICP-OES, germanium
Standardy Standardy kationické Standardy - germanium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.