Tlačiť
Single element standards for ICP-OES, nickel
Dodávateľ: Agilent
5190-8422 5190-8423 5190-8491 5190-8492
HEWL5190-8422EA 155.64 EUR
HEWL5190-8422 HEWL5190-8423 HEWL5190-8491 HEWL5190-8492
Single element standards for ICP-OES, nickel
Standardy Standardy kationické Standardy - nikl
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.