Tlačiť
Tubing
Dodávateľ: VARIAN
AL3000 AL3002 AL3003 R000383005 CP4008 CP4260 5190-6983
VARIAL3000EA 742.88 EUR
VARIAL3000 VARIAL3002 VARIAL3003 VARIR000383005 VARICP4008 VARICP4260 HEWL5190-6983
Tubing
Hadičky Chromatography Tubing
Copper and stainless steel tubing for gas chromatography (GC) applications.
  • Copper can be used for most applications, except those requiring the highest purity gas delivery system
  • Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
These high-quality copper, stainless steel, and UltiMetal Plus tubing are manufactured and cleaned for chromatographic use.