Tlačiť
Single element standards for ICP-OES, Ruthenium
Dodávateľ: Agilent
5190-8442 5190-8443 5190-8513 5190-8514
HEWL5190-8442EA 705.98 EUR
HEWL5190-8442 HEWL5190-8443 HEWL5190-8513 HEWL5190-8514
Single element standards for ICP-OES, Ruthenium
Standardy Standardy kationické Standardy - ruthenium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
These standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.