Tlačiť
Single element standards for ICP-OES, Cadmium
Dodávateľ: Agilent
5190-9414 5190-8328 5190-8366 5190-8367
HEWL5190-9414EA 90.65 EUR
HEWL5190-9414 HEWL5190-8328 HEWL5190-8366 HEWL5190-8367
Single element standards for ICP-OES, Cadmium
Standardy Standardy kationické Standardy - kadmium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
These standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.