Tlačiť
Single element standards for ICP-OES, Sulfur
Dodávateľ: Agilent
5190-8529 5190-8530 5190-8209 5190-8210
HEWL5190-8529EA 95.07 EUR
HEWL5190-8529 HEWL5190-8530 HEWL5190-8209 HEWL5190-8210
Single element standards for ICP-OES, Sulfur
Standardy Standardy kationické Standardy - síra
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
These standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.