Tlačiť
Single element standard for AA and MP-AES, Sodium
Dodávateľ: Agilent
5190-8311 5190-8312
HEWL5190-8311EA 44.93 EUR
HEWL5190-8311 HEWL5190-8312
Single element standard for AA and MP-AES, Sodium
Standardy Standardy kationické Standardy - sodík
Agilent offers a comprehensive range of single and multi-element standards, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.