Tlačiť
Gas line supplies
Dodávateľ: AGILENT HEWLETT PACKARD
0100-2583 G3280-60160 G3280-60163 5064-8092 G3280-67022 G3280-67023 0100-2703
HEWL0100-2583EA 108.3 EUR
HEWL0100-2583 HEWLG3280-60160 HEWLG3280-60163 HEWL5064-8092 HEWLG3280-67022 HEWLG3280-67023 HEWL0100-2703
Gas line supplies
Chromatografické potřeby Chromatography Spare Parts
These gas line supplies add stability and functionality to even the most basic ICP-MS System calibrations.
  • Inner sleeves for carrier, auxiliary and blend gas as well as plasma gas line
  • Argon tubing assembly consisting of 5 m PTFE tubing with one in-line filter and connector set
  • Plasma and auxiliary gas line tubing, Tygon - Int.Ø 3,17 mm/Ext.Ø 6,35 mm/35 cm (for plasma gas line), Int.Ø 2,38 mm/ Ext.Ø 3,96 mm/35 cm (for auxiliary gas line)
  • We provide connection for plasma and auxiliary gas lines to the torch as well as a connector for end cap and gas line
  • Tubing for carrier/makeup gas line, PTFE is provided with Int.Ø 3 mm, Ext.Ø 4 mm, 350 mm
Agilent provide a range of gas line supplies to enable the effective provision of a consistent and stable gas supply to ICP-MS systems. Ensuring stability during basic operation is vitally important for calibrating ICP-MS devices for required levels of sensitivity, a stable gas line helps achieve this. Agilent provide high quality gas line supplies including tubing for a range of lines, inner sleeves and various high quality fittings. To ensure your system's accuracy and longevity replace gas line components in accordance with recommended maintenance schedules.