Tlačiť
Single element standards for ICP-OES, palladium
Dodávateľ: Agilent
5190-8426 5190-8427 5190-8497 5190-8498
HEWL5190-8426EA 840.38 EUR
HEWL5190-8426 HEWL5190-8427 HEWL5190-8497 HEWL5190-8498
Single element standards for ICP-OES, palladium
Standardy Standardy kationické Standardy - palladium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.