Tlačiť
Syringe filters, Captiva
Dodávateľ: AGILENT HEWLETT PACKARD
5190-5307 5190-5308 5190-5309 5190-5277 5190-5278 5190-5279
HEWL5190-5307EA 2085.44 EUR
HEWL5190-5307 HEWL5190-5308 HEWL5190-5309 HEWL5190-5277 HEWL5190-5278 HEWL5190-5279
Syringe filters, Captiva
Filtry Filtry stříkačkové
Syringe filters are used most often to clarify small-volume sample solutions prior to HPLC and ion chromatography.
  • Ideal for busy, high-volume labs
  • Available in a wide range of sizes, formats, and membranes to cover every matrix and sample
  • Excellent flow rates and maximum sample loading capacity
  • Constructed with the highest-grade virgin PP or MBS housing, and is securely welded to prevent bursting and ensure sample integrity
Sample filtration is critical to achieving optimal system performance. Agilent sets the standard for LC/MS testing and certifying more syringe filters to be completely free of observed extractables than any other manufacturer. Premium Syringe Filter Chemical Compatibility Chart Instructions for use - Captiva Syringe Filters For Research Use Only. Not for use in diagnostic procedures.
Informace o dodávce: All Captiva syringe filters are shipped with an HPLC or LC/MS Certificate that guarantees low levels of observed extractables.