Tlačiť
Bio-Monolith HPLC columns
Dodávateľ: AGILENT HEWLETT PACKARD
5069-3639 5069-3637 5069-3636 5069-3635
HEWL5069-3639EA 2435.01 EUR
HEWL5069-3639 HEWL5069-3637 HEWL5069-3636 HEWL5069-3635
Bio-Monolith HPLC columns
Kolony chromatografické
Agilent Bio-Monolith HPLC columns provide high resolution and rapid separations of large biomolecules.
  • Polymer-based, monolith HPLC columns designed for macro biomolecule separations
  • Flow-rate independent separations; no diffusion, no pores and no void volume make transport between mobile and stationary phase very rapid
  • Monolith disk is 5,2×4,95 mm (100 μl column volume) with continuous channels, eliminating diffusion mass transfer
  • Extremely fast separations speed up method development time and decrease costs; locking in method parameters
These HPLC columns provide high resolution and rapid separations of antibodies (IgG, IgM), plasmid DNA, viruses, phages and other macro biomolecules. The product family offers strong cation exchange, strong and weak anion exchange and Protein A phases. Bio-Monolith HPLC columns are compatible with HPLC and preparative LC systems, including Agilent 1100 and 1200 Infinity Series. View our full range of BioHPLC columns for your area of interest, get resources, and take advantage of special offers.