Tlačiť
Single element standards for ICP-OES, holmium
Dodávateľ: Agilent
5190-8465 5190-8466 5190-8396 5190-8397
HEWL5190-8465EA 135.58 EUR
HEWL5190-8465 HEWL5190-8466 HEWL5190-8396 HEWL5190-8397
Single element standards for ICP-OES, holmium
Standardy Standardy kationické Standardy - holmium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.