Tlačiť
Revolve™ Sustainable Sterile Cleanroom Wipes
Dodávateľ: ITW-CONTAMINATION TEXWIPE
STX1704 STX1709 STX1712
115-2707EA 1874.85 EUR
115-2707 115-2709 115-2712
Revolve™ Sustainable Sterile Cleanroom Wipes
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky sterilní do čistých prostor
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment and parts
  • Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
Revolve™ processing provides low levels of ions, NVRs (non volatile residues), particles, and fibres for use in critical cleaning applications and environments. They are designed for high sorption capacity, which is ideal for spill control, cleaning and solution application.
Certifikácie: Meet USP and USP wipe requirements. Sterile wipes are compatible for use in GMP grade A.
Pozor: Post-consumer bottles made from virgin polyester are reused and upcycled through the REPREVE® recycling process to make the yarn used in the Revolve™ products, keeping these bottles from undergoing disposal in landfills.