Tlačiť
TBE Buffer (10X)
Dodávateľ: Apollo
APOSBIA0972-1LEA 74.27 EUR
APOSBIA0972-1L APOSBIA0972-10L APOSBIA0972-5L
TBE Buffer (10X)
Pufry Pufry pro elektroforézu Pufry do dělícího gelu
TBE is employed as an electrophoresis buffer for acrylamide gels and agarose gels for the separation of nucleic acids and, in case of 'simple' applications, for the separation of proteins.
  • Tris-Borate-EDTA buffer
  • 10X concentrated aqueous solution
It is used in the pH range of 8.0 to 8.5.