Tlačiť

Salicylohydroxamic acid

Dodávateľ: Apollo Scientific
Varování

Synonymá: N,2-Dihydroxybenzamide, Salicylhydroxamic acid

OR27156-100G OR27156-25G
APOSOR27156-100GEA 155.02 EUR
APOSOR27156-100G APOSOR27156-25G
Salicylohydroxamic acid
Salicylohydroxamic acid
Vzorec: C₇H₇NO₃
Mw: 153.14 g/mol
Teplota tavenia: 177 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002110
Číslo CAS: 89-73-6
EINECS: 201-934-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR