Tlačiť
VWR®, Cleanroom clogs
Dodávateľ: VWR Collection
114-8980EA 187.86 EUR
114-8980 114-8981 114-8982 114-8983 114-8984 114-8985
VWR®, Cleanroom clogs
Obuv
VWR® cleanroom clogs provide comfort and durability.
  • Anti-static
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non slip sole
  • Anatomic heel basin
They have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day extended comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilisation. They can also be cleaned in 100 °C with domestic detergents, liquid disinfecting solutions, EO, or formaldehyde.
Ideal for use with VWR® cleanroom disposable or launderable socks.
Objednávacie informácie: Clogs can be ordered by the pair or per case.
Balenie: Packed six pairs per case.