Tlačiť
COD photometric test kits, Test’n Tube
Dodávateľ: Hach
2125851 2125951 2415951 2415851 2345952 2345852 2623451
705-0096EA 137.99 EUR
705-0096 705-0097 705-0129 705-0128 HACH2345952 HACH2345852 705-2048
COD photometric test kits, Test’n Tube
Kity pro environmentální testy
Cuvette Test complete with all reagents, optimised for simple handling on the colorimeters DR900 and DR890 and DR850*.
  • Chromsulphuric acid method
Pozor: *705-0096 is not suitable for DR 850
**705-0128 is only suitable for the DR 4000 and the DR 2500