Tlačiť
illustra™ GenomiPhi™ V2 DNA amplification kits
Dodávateľ: Cytiva
25-6600-30 25-6600-31 25-6600-32
25-6600-30EA 273.16 EUR
25-6600-30 25-6600-31 25-6600-32
illustra™ GenomiPhi™ V2 DNA amplification kits
Činidla pro nukleové kyseliny Isothermal Amplification Reagents
illustra™ GenomiPhi™ V2 DNA Amplification Kit, part of the Phi29 DNA polymerase family, is for mini-scale genomic DNA preparation. The protocol is simple. A typical DNA yield of 4 to 7 µg DNA can be achieved in less than two hours with limited hands-on time. The average product length is >10 kb. The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or non purified cell lysate.
  • Quick mini-scale genomic DNA preparation: 4 to 7 μg in <2 hours
  • One simple protocol for all different types of source material
  • Representative amplification of the whole genome
  • High quality DNA for restriction enzyme digestion, hybridisation, cloning, array CGH high throughput genotyping, and DNA archival