Tlačiť
Racks for centrifuge tube cryoboxes
Dodávateľ: TENAK
TE25106 TE25104 TE25100 TE25102
479-0844EA 121.14 EUR
479-0844 479-0843 479-0841 479-0842
Racks for centrifuge tube cryoboxes
Stojany Poličky pro kryoboxy
Stainless steel.
  • Boxes secured with locking rod
  • Can be used with all freezer brands
  • For boxes 130 mm height
For chest freezers. The racks are specially designed for use with boxes for centrifuge tubes, boxes with exterior dimensions up to H×W 132×137 mm.
Objednávacie informácie: Cryoboxes must be ordered separately.