Tlačiť
Joint clamps for spherical ground joints, KECK™
Dodávateľ: DWK LIFE SCIENCES
286340501 286340604 286340707 286340801 286340904
201-7121EA 6.42 EUR
201-7121 201-7122 SCOT286340707 201-7123 201-7124
Joint clamps for spherical ground joints, KECK™
Svorky Svorky pro kónické NZ
POM, coloured. For firm, securely fitting spherical ground joints. These clamps are made of plastic to avoid scratching the glass, as can happen with conventional metal joint clamps.
  • Very good chemical resistance
  • Heat resistant up to 150 °C
  • Sturdy, easy to install and remove