Tlačiť
Eppendorf Xplorer® Electronic Multi-Channel Pipettes
Dodávateľ: EPPENDORF
4861000.104 4861000.112 4861000.120 4861000.139 4861000.147 4861000.155 4861000.163 4861000.171
613-1410EA 1250.7 EUR
613-1410 613-1417 613-1418 613-1419 613-1420 613-1426 613-1427 613-2228
Eppendorf Xplorer® Electronic Multi-Channel Pipettes
Pipety Micropipettes
Eppendorf Xplorer® multi-channel pipettes with dispensing mode provide a cutting-edge solution for high-throughput liquid handling. Offering the highest levels of simplicity, precision, and reproducibility, these pipettes will help you pipette with more speed, comfort, and ergonomics.
  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0,5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker and ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle with powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 and 10 ml)
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Air-cushion pipetting system with epT.I.P.S.® consumables. Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish.
Application: Pipetting of aqueous liquids.
Objednávacie informácie: Supplied with charger.