Tlačiť
Reagent reservoirs, disposable
Dodávateľ: BIOTIX
SR-0025-BWM 63300130 63300133 63300135 63300120 63300121 63300126 63300129 63300124 63300127 63300125 63300128 63300118
613-2857EA 99.48 EUR
613-2857 613-1190 613-1193 613-1191 613-2852 613-2853 BITX63300126 613-2860 BITX63300124 BITX63300127 613-2856 613-2859 613-2850
Reagent reservoirs, disposable
Zásobníky na reagencie
These disposable reagent reservoirs are designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes. Users can choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
  • Sterile or non sterile
  • Angled sides and a V-shaped bottom allow maximum recovery of reagents
  • All reservoirs feature internal graduated volume lines and pour-spouts on the corners
  • Certified free of DNA, DNase, RNase and endotoxins (pyrogens)
Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested assuring maximum quality and compatibility. All raw materials are traceable.
Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.
Balenie: Reagent reservoirs are available pre-sterile, non sterile, bulk and individually wrapped.