Tlačiť
Overhead stirrer tools, anchor stirrers
Dodávateľ: Bohlender
C384-04 C384-02 C384-08 C445-08 C384-64 C384-10 C445-20 C445-34 C384-16 C445-16 C384-06 C445-12 C445-30 C384-17 C384-24 C384-48 C445-44 C445-40 C384-52 C384-44 C384-40 C384-32 C384-28 C384-01 C384-58
441-0415EA 253.91 EUR
441-0415 441-2933 441-0417 441-0077 441-0427 441-0078 441-0086 441-0429 441-0418 441-0075 441-0416 441-0085 441-0428 441-0079 441-0083 441-0423 441-0431 441-0430 441-0425 441-0422 441-0421 441-0420 441-0419 441-0414 441-0426
Overhead stirrer tools, anchor stirrers
Míchadla Nástroje na míchání
PTFE stirrers: These stirrer tools have a PTFE coated stainless steel shaft, and the U-shaped stirrer blade is made entirely of PTFE. Ideal for mixing viscous liquids. Strong, tangential flow with shear rate in the margin area, little sediment on the wall of the vessel.
  • Excellent chemical resistance
  • Temperature resistant from −200 to +250 °C
PTFE blades: Solid PTFE stirrer blades with set of PTFE/PEEK clamp screws, ideal for mixing viscous liquids. Strong, tangential flow with shear rate in the margin area, little sediment on the wall of the vessel.
Solid PTFE stirrer blades with set of PTFE/PEEK clamp screws, ideal for mixing viscous liquids. Strong, tangential flow with shear rate in the margin area, little sediment on the wall of the vessel.
Certifikácie: Conform to FDA requirements.
Informace o dodávce: A spanner is included with the blades for easy assembly.