Tlačiť
UV microplates, 96-well, Corning®
Dodávateľ: Corning
3635 3679
734-1614EA 976.73 EUR
734-1614 734-4120
UV microplates, 96-well, Corning®
Mikrodestičky Mikrodestičky pro HTS
Acrylic with UV transparent flat well bottom, non sterile. Ideal for determining protein and/or nucleic acid concentrations.
  • UV transparent bottom is moulded directly to an acrylic base for greater strength and maximum leak resistance
  • Allows UV absorbance readings with low background, especially at 260 to 280 nm
  • Certified free from DNase and RNase
Objednávacie informácie: Packed 25 per pack, 50 per case.