Tlačiť
96 Well Cluster Tubes, Corning®
Dodávateľ: Corning
4401 4408 4410 4411 4412 4413 4418
732-5554EA 127.16 EUR
732-5554 732-5555 732-5556 732-5557 732-5558 732-5559 732-5560
96 Well Cluster Tubes, Corning®
Zkumavky Zkumavky kultivační
PP tubes, PE tube caps.
  • Composed of 96 PP tubes in a standard microplate format
  • 1,2 ml tubes available individually or in strips of 8 tubes
  • PE tube caps available in 8-cap strips
Balenie: Supplied bagged or racked.