Tlačiť
Corning® Microplate těsnící pásky
Dodávateľ: Corning
6524 6569 6570 3345 6575 4612
734-1756EA 94.53 EUR
734-1756 734-1774 734-1775 732-0486 734-4000 731-0693
Corning® Microplate těsnící pásky
Těsnění Těsnění mikrodestičky
Easy application and removal for short and long-term storage. Provides tight seal to minimise evaporation and condensation.
Aluminium tape: Suitable for use from −80 to +150 °C, not transparent, pierceable, non sterile.
Breathable tape: Allows gas exchange across the surface, sterile.
Universal optical tape: Suitable for use from −70 to +100 °C, transparent, non sterile.
PE sealing tape: Prevents evaporation and enables oil-free operation when used with thermal cyclers with heated lids, non sterile.
Polyester tape: Suitable for use from −40 to +120 °C, transparent, non pierceable, non sterile.