Tlačiť
Safety goggles, 2891
Dodávateľ: 3M
2891-SG 2891S-SG
MMMA2891-SGEA 35.78 EUR
MMMA2891-SG MMMA2891S-SG
Safety goggles, 2891
Brýle bezpečnostní
The 2891 series Safety Goggles are high-wrap, clear polycarbonate safety goggles with Scotchgard™ anti-fog/anti-scratch coating for superior protection.
  • Modern, slim design safety goggles, provide the wearer with an excellent field of vision
  • Anti-fog coating is effective even after multiple washes, disinfecting by soaking in diluted bleach, or using alcohol wipes
  • Wide, nylon elastic strap for simple adjustment and fit
  • Clear lens for excellent protection against Ultraviolet (UV) radiation
  • Strong polycarbonate lens for impact resistance
  • Lens marking: 2C-1.2 3M 1 BT 9 KN CE
  • Frame marking: 3M EN166 3459 BT CE 3M 2891S
For eye protection with an excellent field of vision, protection from gases and vapours, and reduced fogging in hot or humid conditions, use the 2890 series safety goggles. The modern, slim design features a clear, strong polycarbonate lens for impact resistance. The wide, nylon elastic strap provides simple adjustment to ensure a close and comfortable fit.
Goggles are available with a sealed design to help provide protection against liquids, dust, gases, and vapours, or with indirect ventilation to improve air circulation.
3M Scotchgard™ anti-fog coating helps ensure clear visibility, resisting fogging longer than traditional anti-fog coatings even after washing multiple times, including using alcohol wipes or disinfecting by soaking in diluted bleach. Superior anti-fog and anti-scratch properties provide increased durability, visibility, and meet K&N marking requirements of EN166.
Certifikácie: EN 166