Tlačiť
Electronic contact thermometers, GFX 460
Dodávateľ: EBRO
1340-5460 1340-5464
620-1386EA 440.62 EUR
620-1386 471-1060
Electronic contact thermometers, GFX 460
Teploměry Teploměry digitální
These electronic contact thermometers are suitable for applications with all magnetic stirrers.
  • High accuracy through fuzzy logic
  • Minimises exceeding of temperature limit
  • Status shown on LED illuminated displays
DIN 45322 (diode connection). Safety circuit according to DIN 12878 class 1 and 2.
Certifikácie: DIN 45322 (diode connection). Safety circuit according to DIN 12878 class 1 and 2.
Informace o dodávce: Supplied with external 9 to 12 V DC, 20 mA power supply.