Tlačiť

Platinum(II) acetylacetonate, Sigma-Aldrich®

Dodávateľ: Merck
Varování

Synonymá: Platinum (II) bis(2,4-pentanedionato-O,O'), Platinum (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')platinum (II)

M-Clarity™ quality level = MQ100

282782-500MG 282782-5G
SIAL282782-500MGEA 347.69 EUR
SIAL282782-500MG SIAL282782-5G
Platinum(II) acetylacetonate, Sigma-Aldrich®
Platinum(II) acetylacetonate
Vzorec: C₁₀H₁₄O₄Pt
Mw: 393.3
Teplota tavenia: 250…252 °C
Hustota: ∼1 g/cm³ (20 °C)
Číslo MDL: MFCD00000028
Číslo CAS: 15170-57-7
EINECS: 239-223-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR