Disoprofol ≥97%
Dodávateľ: Thermo Fisher Scientific
|
Varování
|
Synonymá:
Propofol, 1,3-Di(methylethyl)-2-hydroxybenzene, 2,6-Diisopropylphenol, 2,6-Bisisopropylphenol
Purity: 97%
CAS No.: 2078-54-8
Molecular Formula: C12H18O
Formula Weight: 178.28
Flash Point: 113°(235°F)
Density: 0.962
Refractive Index: 1.5140
Melting Point: 17-18°
Boiling Point: 254-256°
Beilstein Reference: 1866484
Merck Reference: 14,7834
MDL No.: MFCD00008885
An anesthetic that exhibits direct activation of the GABA-A receptor
Vzorec:
C₁₂H₁₈O Teplota varu: 254…256 °C Teplota tavenia: 17…18 °C Hustota: 0,962 Teplota vzplanutia: 113 °C (235 °F) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00008885 Číslo CAS: 2078-54-8 EINECS: 218-206-6 Merck Index: 14,7834 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...