Tlačiť

8-Aminocaprylic acid, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid

M-Clarity™ quality level = MQ200

855294-1G 855294-5G
SIAL855294-1GEA 83.72 EUR
SIAL855294-1G SIAL855294-5G
8-Aminocaprylic acid, Sigma-Aldrich®
8-Aminocaprylic acid
Vzorec: C₈H₁₇NO₂
Mw: 159.23
Teplota varu: 289
Teplota tavenia: 192…192.5 °C
Hustota: 1.221 g/cm³ (20 °C)
Číslo MDL: MFCD00008245
Číslo CAS: 1002-57-9
EINECS: 213-687-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR