Tlačiť
Tapes, double coated, GPT-020F, 3M
Dodávateľ: 3M
GPT-020F-100MM GPT-020F-12MM GPT-020F-19MM GPT-020F-25MM GPT-020F-50MM
MMMAGPT-020F-100MMEA 644.2 EUR
MMMAGPT-020F-100MM MMMAGPT-020F-12MM MMMAGPT-020F-19MM MMMAGPT-020F-25MM MMMAGPT-020F-50MM
Tapes, double coated, GPT-020F, 3M
Pásky
Designed for versatility, 3M™ double coated GPT-020F tape is a transparent, 200 µm thick, modified solventless acrylic adhesive with a white printed PP filmic liner. This tape combines a high level of peel and shear performance, enabling high adhesion to a wide variety of substrates both to high and some low surafce energy substrates.
  • Providing versatility with purpose
  • High initial tack and humidity resistance
  • Easy handling and converting due to filmic liner delivery
  • High temperature resistance, providing full bonding performance, up to 90 °C
  • Self-adhesive mounting of furniture trim, sealing profiles and cable ducts
  • Length: 50 m
3M™ double coated GPT-020F tape offers high initial tack and humidity resistance, providing easy handling and converting due to the filmic liner delivery.
Application includes bonding and mounting of sales displays and billboards; fixing of decorative trims and emblems.
Store at 16 to 25 °C and 40 to 65% relative humidity in its original box.