Tlačiť
VWR®, HPTLC Glass/Aluminium Plates, Silica, Modified Layers
Dodávateľ: VWR Collection
551-0029EA 515.84 EUR
551-0029 551-0030
VWR®, HPTLC Glass/Aluminium Plates, Silica, Modified Layers
TLC desky
Modified silica layers are, depending on the modification, suitable for polar to unpolar eluents or eluent mixtures.
  • RP-18w is a special modified sorbent which is water wettable
Indicator: Acid-resistant with a pale blue fluorescence for short wavelength UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 2 to 10 µm