Tlačiť
VWR®, HPTLC Glass Plates, Silica HP, Unmodified Layers
Dodávateľ: VWR Collection
551-0009EA 493.78 EUR
551-0009 551-0023 551-0027 551-0028
VWR®, HPTLC Glass Plates, Silica HP, Unmodified Layers
TLC desky
These HPTLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.
  • Specific dyeability and abrasion resistance based on an optimised binder system
  • Good wettability for exact colorisation results, even with 100% aqueous detection reagents
Specific surface (BET): ~500 m²/g
Specific pore volume: 0,75 ml/g
Particle size: 2 to 10 µm