Tlačiť
Precision balances, Entris® II Basic Essential series
Dodávateľ: Sartorius Balances
BCE1202-1S BCE1202I-1CEU BCE1202I-1S BCE2201-1S BCE2201I-1CEU BCE2201I-1S BCE2202-1S BCE2202I-1CEU BCE2202I-1S BCE223-1S BCE223I-1CEU BCE223I-1S BCE3202-1S BCE3202I-1CEU BCE3202I-1S BCE323-1S BCE323I-1CEU BCE323I-1S BCE4202-1S BCE4202I-1CEU BCE4202I-1S BCE423-1S BCE423I-1CEU BCE423I-1S BCE5201-1S BCE5201I-1CEU BCE5201I-1S BCE6200-1S BCE6200I-1CEU BCE6200I-1S BCE6202-1S BCE6202I-1CEU BCE6202I-1S BCE622-1S BCE622I-1CEU BCE622I-1S BCE623-1S BCE623I-1CEU BCE623I-1S BCE653-1S BCE653I-1CEU BCE653I-1S BCE8200-1S BCE8200I-1CEU BCE8200I-1S BCE8201-1S BCE8201I-1CEU BCE8201I-1S BCE822-1S BCE822I-1CEU BCE822I-1S
611-4560EA 1597.67 EUR
611-4560 611-4566 611-4568 611-4578 611-4579 611-4586 611-4587 611-4588 611-4590 611-4598 611-4599 611-4605 611-4610 611-4611 611-4613 611-4614 611-4615 611-4617 611-4618 611-4619 611-4621 611-4622 611-4623 611-4625 611-4626 611-4627 611-4629 611-4630 611-4631 611-4633 611-4634 611-4635 611-4637 611-4638 611-4639 611-4641 611-4642 611-4643 611-4645 611-4650 611-4651 611-4653 611-4654 611-4655 611-4657 611-4658 611-4659 611-4661 611-4662 611-4663 611-4665
Precision balances, Entris® II Basic Essential series
Váhy
Entris® II Basic Essential precision balances feature isoCAL, LED touch technology screen and 12 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard-wearing PBT, stainless steel and glass.
  • Fast stabilisation time thanks to state-of-the-art weighing sensors
  • Best repeatability with rectangular weighing pan
  • Overload protection, rugged design, weighs up to a pre-set amount
  • Guaranteed reliability with the self-test ‘@start’
  • Choice of models with internal or external calibration
  • Hybrid screen gives excellent readability and use. Combines intuitive, wear resistant LED and touch technology provides an easy and clear structured user interface
  • Password protection ensures secure operation, avoids unintentional changes
Only the 3 decimal place models have a draft shield, this minimises weighing errors caused by electrostatically charged samples using specially coated glass parts. In models up to 620 g the weighing chamber gives the highest usable weighing chamber height easily accommodating even large containers, ensuring convenient access to the weighing pan. The sliding and top doors can be can easily be removed for cleaning. The 650 g models have smaller, round draft shields.
Choice of models with isoCAL (internal calibration and adjustment) for accurate weighing results. Optimal accuracy and operating convenience using fully automated internal temperature and time-controlled calibration and adjustment feature. Assures (SOP) compliant operation with self-notification if calibration is outside the normal range. The balance easily adapts to the ambient conditions with just one click on the home screen.
'Plug-and-play' convenience - automatically detects Sartorius accessories (e.g. printer, second display). Real 'PC-direct feature' for easy connection to a PC to transfer weighing data directly into spreadsheets or documents such as Microsoft® Excel or Word.
Entris® II Basic Essential balances have 12 built-in applications with GLP/GMP compliant printout/data output. All models have 8 built-in languages for international operation: English, German, French, Italian, Spanish, Portuguese, Russian and Polish. The series has data output for dynamic weighing applications and configurable time interval for data output. Modern, up-to-date connectivity methods, USB Type C interface, RS232 9-pin interface and a RS232 port.
Pozor: Models with suffix -CEU are verified version Europe (except France, where -CFR models are available).