Tlačiť
Worktables
Dodávateľ: HUPFER METALLWERKE
7504583 75R60302-1800 75R60302-2000
132-0017EA 3711.59 EUR
132-0017 HUPF75R60302-1800 HUPF75R60302-2000
Worktables
Nábytek kancelářský Stoly
This open version work table features closed table top and bottom side covered with 0,8 mm sheet steel.
  • Particle board stuffing
  • Sandwich panelled table top
Working table made of stainless steel with grain size 400 ground and material thickness of 1 mm.