Tlačiť
VWR®, Spare Caps for VWR® Carboys
Dodávateľ: VWR Collection
216-1840EA 16.85 EUR
216-1840 216-1841
VWR®, Spare Caps for VWR® Carboys
Uzávěry Uzávěry k láhvím
Spare caps are designed for use with either narrow mouth or wide mouth VWR® carboys
Autoclavable.