Tlačiť
Flash point standards, Cleveland open cup, ASTM D 92, Supelco®
Dodávateľ: Merck
FPCOC5-3X80ML FPCOC1-3X80ML FPCOC3-3X80ML FPCOC4-3X80ML FPCOC6-3X80ML
SIALFPCOC5-3X80MLEA 349.78 EUR
SIALFPCOC5-3X80ML SIALFPCOC1-3X80ML SIALFPCOC3-3X80ML SIALFPCOC4-3X80ML SIALFPCOC6-3X80ML
Flash point standards, Cleveland open cup, ASTM D 92, Supelco®
Standardy Standardy pro ropný průmysl
These UKAS certified standards are specifically designed to be used for regular instrument performance checks.
  • Mineral oils
Certifikácie: These products are certified reference materials (CRMs) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.